Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01435086_g1
          See other SERPINB6B GE Assays ›
          Gene Symbol:
          Serpinb6b
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade B, member 6b
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 32770488 - 32787835 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01435086_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpinb6b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012214.1 NP_001012214.1 6 - 7 768 117
          GenBank mRNA BC079108.1 - 6 - 7 768 7125468 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001669.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpinb6b

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P683837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56271 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76186 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P683837 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P683837 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P683837 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P683837 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51112 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76181 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10798145 - -
          Affymetrix Rat Gene 1.0 ST Array 10798146 - -
          Affymetrix Rat Gene 1.0 ST Array 10798149 - -
          Affymetrix Rat Gene 1.0 ST Array 10798143 - -
          Affymetrix Rat Gene 1.0 ST Array 10798144 - -
          Affymetrix Rat Gene 1.1 ST Array 10798142 - -
          Rat Genome 230 2.0 Array 1397866_at - -
          Rat Genome 230 2.0 Array 1397866_PM_at - -
          Affymetrix HT Rat Focus Array 1397866_PM_at - -
          Affymetrix HT Rat Focus Array 1397866_at - -
          Affymetrix Rat Clariom S Assay PSR1700011575.rn.1 TC1700001669.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700011576.rn.1 TC1700001669.rn.1 RTA TC
          Rat Expression Set 230 1397866_at - -
          Rat Expression Set 230 1397866_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10798142 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376788 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375624 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline