Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01435110_m1
          See other PRL7B1 GE Assays ›
          Gene Symbol:
          Prl7b1
          Gene Name:
          prolactin family 7, subfamily b, member 1
          Gene Aliases:
          Prlpn
          Chromosome Location:
          Chr.17: 39153431 - 39161644 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01435110_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prl7b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153738.1 NP_714960.1 2 - 3 237 133
          GenBank mRNA AF525159.1 - 2 - 3 237 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000592.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prl7b1

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Process(es)

          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P049301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472919 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33797 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049301 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049301 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P049301 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P132628 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19795 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10795015 - -
          Affymetrix Rat Gene 1.0 ST Array 10795020 - -
          Affymetrix Rat Gene 1.0 ST Array 10795019 - -
          Affymetrix Rat Gene 1.0 ST Array 10795016 - -
          Affymetrix Rat Gene 1.0 ST Array 10795018 - -
          Affymetrix Rat Gene 1.0 ST Array 10795014 - -
          Affymetrix Rat Gene 1.0 ST Array 10795017 - -
          Affymetrix Rat Gene 1.1 ST Array 10795013 - -
          Rat Genome 230 2.0 Array 1378483_PM_at - -
          Rat Genome 230 2.0 Array 1378483_at - -
          Affymetrix HT Rat Focus Array 1378483_PM_at - -
          Affymetrix HT Rat Focus Array 1378483_at - -
          Affymetrix Rat Clariom S Assay PSR1700004062.rn.1 TC1700000592.rn.1 RTA TC
          Rat Expression Set 230 1378483_PM_at - -
          Rat Expression Set 230 1378483_at - -
          Affymetrix Rat Gene 1.0 ST Array 10795013 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21660360 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369152 - -
          RatRef-12 Expression BeadChip ILMN_57645 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline