Search
Search

Snrnp48| Gene Symbol: Snrnp48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106107.1 | NP_001099577.1 | 6 - 7 | 747 | 89 | |
| XM_006253787.2 | XP_006253849.1 | 6 - 7 | 769 | 89 | ||
| XM_006253788.2 | XP_006253850.1 | 6 - 7 | 769 | 89 | ||
| GenBank mRNA | BC088167.1 | - | 6 - 7 | 738 | 89 | |
Set Membership: |
|