Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01435293_m1
          See other SPTLC1 GE Assays ›
          Gene Symbol:
          Sptlc1
          Gene Name:
          serine palmitoyltransferase, long chain base subunit 1
          Gene Aliases:
          Lcb1, RGD1306617, Spt1
          Chromosome Location:
          Chr.17: 11856525 - 11895566 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01435293_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sptlc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108406.1 NP_001101876.1 14 - 15 1334 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000208.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          transaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          sphingomyelin biosynthetic process
          sphinganine biosynthetic process
          sphingosine biosynthetic process
          ceramide biosynthetic process
          serine C-palmitoyltransferase activity
          pyridoxal phosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P734384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170616 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106669 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109580 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P734384 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P734384 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P734384 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P734384 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P314081 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P440870 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104725 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10794220 - -
          Affymetrix Rat Gene 1.0 ST Array 10794211 - -
          Affymetrix Rat Gene 1.0 ST Array 10794212 - -
          Affymetrix Rat Gene 1.0 ST Array 10794219 - -
          Affymetrix Rat Gene 1.0 ST Array 10794213 - -
          Affymetrix Rat Gene 1.0 ST Array 10794217 - -
          Affymetrix Rat Gene 1.0 ST Array 10794210 - -
          Affymetrix Rat Gene 1.0 ST Array 10794221 - -
          Affymetrix Rat Gene 1.0 ST Array 10794208 - -
          Affymetrix Rat Gene 1.0 ST Array 10794215 - -
          Affymetrix Rat Gene 1.0 ST Array 10794207 - -
          Affymetrix Rat Gene 1.0 ST Array 10794216 - -
          Affymetrix Rat Gene 1.0 ST Array 10794218 - -
          Affymetrix Rat Gene 1.0 ST Array 10794209 - -
          Affymetrix Rat Gene 1.0 ST Array 10794214 - -
          Affymetrix Rat Gene 1.1 ST Array 10794206 - -
          Rat Genome 230 2.0 Array 1396024_PM_at - -
          Rat Genome 230 2.0 Array 1373203_at - -
          Rat Genome 230 2.0 Array 1373203_PM_at - -
          Rat Genome 230 2.0 Array 1396024_at - -
          Affymetrix HT Rat Focus Array 1396024_at - -
          Affymetrix HT Rat Focus Array 1396024_PM_at - -
          Affymetrix HT Rat Focus Array 1373203_PM_at - -
          Affymetrix HT Rat Focus Array 1373203_at - -
          Affymetrix Rat Clariom S Assay PSR1700001719.rn.1 TC1700000208.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001718.rn.1 TC1700000208.rn.1 RTA TC
          Rat Expression Set 230 1373203_at - -
          Rat Expression Set 230 1373203_PM_at - -
          Rat Expression Set 230 1396024_PM_at - -
          Rat Expression Set 230 1396024_at - -
          Affymetrix Rat Gene 1.0 ST Array 10794206 - -
          Rat Genome U34 Set rc_AA892380_at - -
          Rat Genome U34 Set rc_AI105284_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21585486 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365055 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline