Search
Search
| Gene Symbol: Abca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024396.1 | NP_077372.1 | 9 - 10 | 1489 | 94 | |
| XM_006233645.2 | XP_006233707.1 | 10 - 11 | 1598 | 94 | ||
| XM_008761609.1 | XP_008759831.1 | 9 - 10 | 4165 | 94 | ||
| GenBank mRNA | AB037937.1 | - | 9 - 10 | 1489 | 94 | |
Set Membership: |
|