Search
Search
| Gene Symbol: Fubp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039337.1 | NP_001034426.1 | 11 - 12 | 976 | 60 | |
| XM_006233921.2 | XP_006233983.1 | 11 - 12 | 1121 | 60 | ||
| XM_006233922.2 | XP_006233984.1 | 11 - 12 | 1121 | 60 | ||
| XM_006233924.2 | XP_006233986.1 | 11 - 12 | 1122 | 60 | ||
| GenBank mRNA | DQ144645.1 | - | 11 - 12 | 976 | 60 | |
| FQ212354.1 | - | 11 - 12 | 1054 | 60 | ||
Set Membership: |
|