Search
Search
| Gene Symbol: Sptan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171983.2 | NP_741984.2 | 8 - 9 | 1193 | 59 | |
| XM_008761673.1 | XP_008759895.1 | 8 - 9 | 1193 | 59 | ||
| XM_008761674.1 | XP_008759896.1 | 8 - 9 | 1193 | 59 | ||
| XM_008761675.1 | XP_008759897.1 | 8 - 9 | 1193 | 59 | ||
| XM_008761676.1 | XP_008759898.1 | 8 - 9 | 1193 | 59 | ||
| XM_008761677.1 | XP_008759899.1 | 8 - 9 | 1193 | 59 | ||
| XM_008761678.1 | XP_008759900.1 | 8 - 9 | 1193 | 59 | ||
| GenBank mRNA | AF084186.1 | - | 7 - 8 | 1085 | 59 | |
| BC070885.1 | - | 8 - 9 | 1157 | 7098134 | 59 | |
| X90845.1 | - | 8 - 9 | 1193 | 59 | ||
Set Membership: |
|