Search
Search
| Gene Symbol: Gtf3c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079941.1 | NP_001073410.1 | 3 - 4 | 822 | 87 | |
| XM_006233835.2 | XP_006233897.1 | 3 - 4 | 831 | 87 | ||
| GenBank mRNA | BC129109.1 | - | 3 - 4 | 822 | 7105121 | 87 |
Set Membership: |
|