Search
Search
| Gene Symbol: Ralgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019250.2 | NP_062123.2 | 9 - 10 | 1753 | 71 | |
| XM_006233750.2 | XP_006233812.1 | 9 - 10 | 2298 | 71 | ||
| XM_006233751.2 | XP_006233813.1 | 9 - 10 | 4231 | 71 | ||
| XM_006233752.2 | XP_006233814.1 | 9 - 10 | 1811 | 71 | ||
| XM_006233753.2 | XP_006233815.1 | 9 - 10 | 1775 | 71 | ||
| XM_008761632.1 | XP_008759854.1 | 9 - 10 | 2076 | 71 | ||
| GenBank mRNA | L07925.1 | - | 9 - 10 | 1753 | 71 | |
Set Membership: |
|