Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01436557_m1
          See other GRHPR GE Assays ›
          Gene Symbol:
          Grhpr
          Gene Name:
          glyoxylate reductase/hydroxypyruvate reductase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 60528981 - 60538410 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01436557_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grhpr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113754.1 NP_001107226.1 8 - 9 927 53
          GenBank mRNA BC158680.1 - 8 - 9 911 8373346 53

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000561.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          excretion
          dicarboxylic acid metabolic process
          protein oligomerization
          oxidation-reduction process
          glycerate dehydrogenase activity
          hydroxypyruvate reductase activity
          glyoxylate reductase (NADP) activity
          carboxylic acid binding
          protein homodimerization activity
          NADP binding
          NAD binding
          NADPH binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157313 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P141510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504625 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55684 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157313 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141510 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141510 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157313 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157313 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P141510 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P140449 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10868680 - -
          Affymetrix Rat Gene 1.0 ST Array 10868678 - -
          Affymetrix Rat Gene 1.0 ST Array 10868681 - -
          Affymetrix Rat Gene 1.0 ST Array 10868675 - -
          Affymetrix Rat Gene 1.0 ST Array 10868676 - -
          Affymetrix Rat Gene 1.0 ST Array 10868677 - -
          Affymetrix Rat Gene 1.0 ST Array 10868674 - -
          Affymetrix Rat Gene 1.0 ST Array 10868679 - -
          Affymetrix Rat Gene 1.1 ST Array 10868673 - -
          Rat Genome 230 2.0 Array 1373337_at - -
          Rat Genome 230 2.0 Array 1373337_PM_at - -
          Affymetrix HT Rat Focus Array 1373337_PM_at - -
          Affymetrix HT Rat Focus Array 1373337_at - -
          Affymetrix Rat Clariom S Assay PSR0500003643.rn.1 TC0500000561.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500003640.rn.1 TC0500000561.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500003641.rn.1 TC0500000561.rn.1 RTA TC
          Rat Expression Set 230 1373337_PM_at - -
          Rat Expression Set 230 1373337_at - -
          Affymetrix Rat Gene 1.0 ST Array 10868673 - -
          Rat Genome U34 Set rc_AA892799_r_at - -
          Rat Genome U34 Set rc_AA892799_s_at - -
          Rat Genome U34 Set rc_AA892799_i_at - -
          Rat Genome U34 Set rc_AA924630_g_at - -
          Rat Genome U34 Set rc_AA924630_at - -
          Rat Genome U34 Set H31117_at - -
          Rat Genome U34 Set rc_AI232066_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20715773 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline