Search
Search
| Gene Symbol: Ankrd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134969.1 | NP_001128441.1 | 13 - 14 | 1722 | 64 | |
| XM_006237976.2 | XP_006238038.1 | 13 - 14 | 1651 | 64 | ||
| XM_006237977.2 | XP_006238039.1 | 13 - 14 | 1623 | 64 | ||
| XM_006237978.2 | XP_006238040.1 | 13 - 14 | 2081 | 64 | ||
| XM_006237981.2 | XP_006238043.1 | 9 - 10 | 1077 | 64 | ||
| XM_008763638.1 | XP_008761860.1 | 13 - 14 | 2081 | 64 | ||
| XM_008763639.1 | XP_008761861.1 | 12 - 13 | 1524 | 64 | ||
| XM_008763640.1 | XP_008761862.1 | 13 - 14 | 1623 | 64 | ||
| GenBank mRNA | BC168675.1 | - | 14 - 15 | 1658 | 7939387 | 64 |
Set Membership: |
|