Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01436790_m1
          See other LRRC69 GE Assays ›
          Gene Symbol:
          Lrrc69
          Gene Name:
          leucine rich repeat containing 69
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 28133068 - 28186803 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01436790_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134624.1 NP_001128096.1 195 150

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002345.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P560428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841904 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89380 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156979 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12355 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120582 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169297 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41711 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45505 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56067 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119560 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22738 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58082 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167453 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139981 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29475 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P560428 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P560428 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P560428 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P560428 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10875606 - -
          Affymetrix Rat Gene 1.0 ST Array 10875608 - -
          Affymetrix Rat Gene 1.0 ST Array 10875609 - -
          Affymetrix Rat Gene 1.0 ST Array 10875607 - -
          Affymetrix Rat Gene 1.0 ST Array 10875605 - -
          Affymetrix Rat Gene 1.0 ST Array 10875603 - -
          Affymetrix Rat Gene 1.0 ST Array 10875604 - -
          Affymetrix Rat Gene 1.1 ST Array 10875601 - -
          Affymetrix Rat Clariom S Assay PSR0500017703.rn.1 TC0500002345.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017706.rn.1 TC0500002345.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017714.rn.1 TC0500002345.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017715.rn.1 TC0500002345.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017716.rn.1 TC0500002345.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017702.rn.1 TC0500002345.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017709.rn.1 TC0500002345.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017712.rn.1 TC0500002345.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017713.rn.1 TC0500002345.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017711.rn.1 TC0500002345.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10875601 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22106521 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline