Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01436887_m1
          See other RIPK2 GE Assays ›
          Gene Symbol:
          Ripk2
          Gene Name:
          receptor-interacting serine-threonine kinase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 29839477 - 29870390 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01436887_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ripk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191865.1 NP_001178794.1 8 - 9 1159 125
          GenBank mRNA BC107468.1 - 6 - 7 590 125
          FQ230093.1 - 2 - 3 126 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002363.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytokine-mediated signaling pathway
          adaptive immune response
          positive regulation of T-helper 1 type immune response
          protein phosphorylation
          I-kappaB kinase/NF-kappaB signaling
          positive regulation of peptidyl-threonine phosphorylation
          positive regulation of protein ubiquitination
          lipopolysaccharide-mediated signaling pathway
          positive regulation of chemokine production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-2 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          positive regulation of stress-activated MAPK cascade
          positive regulation of immature T cell proliferation
          positive regulation of peptidyl-serine phosphorylation
          toll-like receptor 2 signaling pathway
          toll-like receptor 4 signaling pathway
          T cell proliferation
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to exogenous dsRNA
          innate immune response
          positive regulation of T-helper 1 cell differentiation
          positive regulation of JNK cascade
          positive regulation of alpha-beta T cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          defense response to Gram-positive bacterium
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of ERK1 and ERK2 cascade
          nucleotide-binding oligomerization domain containing 1 signaling pathway
          nucleotide-binding oligomerization domain containing 2 signaling pathway
          response to interleukin-1
          response to interleukin-12
          response to interleukin-18
          cellular response to lipopolysaccharide
          cellular response to lipoteichoic acid
          cellular response to peptidoglycan
          cellular response to muramyl dipeptide
          protein serine/threonine kinase activity
          ATP binding
          LIM domain binding
          protein homodimerization activity
          CARD domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P811606 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P811606 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P811606 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P811606 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18344 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P811606 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85187 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10875657 - -
          Affymetrix Rat Gene 1.0 ST Array 10875662 - -
          Affymetrix Rat Gene 1.0 ST Array 10875660 - -
          Affymetrix Rat Gene 1.0 ST Array 10875664 - -
          Affymetrix Rat Gene 1.0 ST Array 10875658 - -
          Affymetrix Rat Gene 1.0 ST Array 10875663 - -
          Affymetrix Rat Gene 1.0 ST Array 10875659 - -
          Affymetrix Rat Gene 1.0 ST Array 10875661 - -
          Affymetrix Rat Gene 1.1 ST Array 10875656 - -
          Rat Genome 230 2.0 Array 1381279_PM_at - -
          Rat Genome 230 2.0 Array 1381279_at - -
          Affymetrix HT Rat Focus Array 1381279_PM_at - -
          Affymetrix HT Rat Focus Array 1381279_at - -
          Affymetrix Rat Clariom S Assay PSR0500017820.rn.1 TC0500002363.rn.1 RTA TC
          Rat Expression Set 230 1381279_at - -
          Rat Expression Set 230 1381279_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10875656 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21577647 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365904 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352868 - -
          RatRef-12 Expression BeadChip ILMN_59534 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0