Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01436954_m1
          See other CORO2A GE Assays ›
          Gene Symbol:
          Coro2a
          Gene Name:
          coronin, actin binding protein 2A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 62154526 - 62212554 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01436954_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Coro2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012101.2 NP_001012101.2 9 - 10 1299 63
          XM_006238058.2 XP_006238120.1 8 - 9 2854 63
          GenBank mRNA BC079209.1 - 9 - 10 1252 7131801 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002689.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Coro2a

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          actin cytoskeleton organization
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P163944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171302 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101442 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87658 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146568 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67570 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P163944 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P163944 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P163944 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P163944 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10259 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29058 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46494 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21310 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10876604 - -
          Affymetrix Rat Gene 1.0 ST Array 10876610 - -
          Affymetrix Rat Gene 1.0 ST Array 10876614 - -
          Affymetrix Rat Gene 1.0 ST Array 10876608 - -
          Affymetrix Rat Gene 1.0 ST Array 10876602 - -
          Affymetrix Rat Gene 1.0 ST Array 10876612 - -
          Affymetrix Rat Gene 1.0 ST Array 10876603 - -
          Affymetrix Rat Gene 1.0 ST Array 10876609 - -
          Affymetrix Rat Gene 1.0 ST Array 10876607 - -
          Affymetrix Rat Gene 1.0 ST Array 10876611 - -
          Affymetrix Rat Gene 1.0 ST Array 10876613 - -
          Affymetrix Rat Gene 1.0 ST Array 10876606 - -
          Affymetrix Rat Gene 1.0 ST Array 10876605 - -
          Affymetrix Rat Gene 1.0 ST Array 10876615 - -
          Affymetrix Rat Gene 1.1 ST Array 10876601 - -
          Rat Genome 230 2.0 Array 1379656_PM_a_at - -
          Rat Genome 230 2.0 Array 1380960_at - -
          Rat Genome 230 2.0 Array 1379656_a_at - -
          Rat Genome 230 2.0 Array 1380960_PM_at - -
          Affymetrix HT Rat Focus Array 1380960_at - -
          Affymetrix HT Rat Focus Array 1380960_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500020037.rn.1 TC0500002689.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500020036.rn.1 TC0500002689.rn.1 RTA TC
          Rat Expression Set 230 1380960_PM_at - -
          Rat Expression Set 230 1380960_at - -
          Affymetrix Rat Gene 1.0 ST Array 10876601 - -
          Rat Genome U34 Set rc_AA926011_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22069094 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376577 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375888 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline