Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01437032_g1
          See other FAM221B GE Assays ›
          Gene Symbol:
          Fam221b
          Gene Name:
          family with sequence similarity 221, member B
          Gene Aliases:
          RGD1310425
          Chromosome Location:
          Chr.5: 59156079 - 59165250 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01437032_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam221b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013934.1 NP_001013956.1 6 - 7 1639 62
          XM_008763669.1 XP_008761891.1 6 - 7 1864 62
          XM_008763670.1 XP_008761892.1 6 - 7 1849 62
          GenBank mRNA BC081906.1 - 6 - 7 1639 7111080 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500004256.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam221b

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P314246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175529 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P314246 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P314246 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P314246 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P314246 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1163 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34996 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10876444 - -
          Affymetrix Rat Gene 1.0 ST Array 10876445 - -
          Affymetrix Rat Gene 1.0 ST Array 10876449 - -
          Affymetrix Rat Gene 1.0 ST Array 10876447 - -
          Affymetrix Rat Gene 1.0 ST Array 10876443 - -
          Affymetrix Rat Gene 1.0 ST Array 10876450 - -
          Affymetrix Rat Gene 1.0 ST Array 10876441 - -
          Affymetrix Rat Gene 1.0 ST Array 10876442 - -
          Affymetrix Rat Gene 1.0 ST Array 10876448 - -
          Affymetrix Rat Gene 1.1 ST Array 10876440 - -
          Rat Genome 230 2.0 Array 1385288_at - -
          Rat Genome 230 2.0 Array 1385288_PM_at - -
          Affymetrix HT Rat Focus Array 1385288_PM_at - -
          Affymetrix HT Rat Focus Array 1385288_at - -
          Affymetrix Rat Clariom S Assay PSR0500032963.rn.1 TC0500004256.rn.1 RTA TC
          Rat Expression Set 230 1385288_at - -
          Rat Expression Set 230 1385288_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10876440 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21493473 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369713 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline