Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01437268_m1
          See other PMS1 GE Assays ›
          Gene Symbol:
          Pms1
          Gene Name:
          postmeiotic segregation increased 1 (S. cerevisiae)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 53120656 - 53206520 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01437268_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pms1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009535.1 NP_001009535.1 9 - 10 1906 119
          GenBank mRNA BC061722.1 - 9 - 10 1906 5598508 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000606.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pms1

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mismatch repair
          response to drug
          molecular_function
          single-stranded DNA binding
          ATP binding
          ATPase activity
          mismatched DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P155716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86720 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157801 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108505 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54178 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3726 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145195 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169789 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67198 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155716 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155716 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P155716 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17960 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97933 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82336 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22971 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28629 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4155 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60109 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96050 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38405 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10923166 - -
          Affymetrix Rat Gene 1.0 ST Array 10923175 - -
          Affymetrix Rat Gene 1.0 ST Array 10923164 - -
          Affymetrix Rat Gene 1.0 ST Array 10923172 - -
          Affymetrix Rat Gene 1.0 ST Array 10923165 - -
          Affymetrix Rat Gene 1.0 ST Array 10914848 - -
          Affymetrix Rat Gene 1.0 ST Array 10923168 - -
          Affymetrix Rat Gene 1.0 ST Array 10923167 - -
          Affymetrix Rat Gene 1.0 ST Array 10923170 - -
          Affymetrix Rat Gene 1.0 ST Array 10923176 - -
          Affymetrix Rat Gene 1.0 ST Array 10923169 - -
          Affymetrix Rat Gene 1.0 ST Array 10923163 - -
          Affymetrix Rat Gene 1.0 ST Array 10923162 - -
          Affymetrix Rat Gene 1.0 ST Array 10923173 - -
          Affymetrix Rat Gene 1.1 ST Array 10914844 - -
          Affymetrix Rat Gene 1.1 ST Array 10923161 - -
          Rat Genome 230 2.0 Array 1384523_at - -
          Rat Genome 230 2.0 Array 1384523_PM_at - -
          Affymetrix HT Rat Focus Array 1384523_PM_at - -
          Affymetrix HT Rat Focus Array 1384523_at - -
          Affymetrix Rat Clariom S Assay PSR0900004942.rn.1 TC0900000606.rn.1 RTA TC
          Rat Expression Set 230 1384523_PM_at - -
          Rat Expression Set 230 1384523_at - -
          Affymetrix Rat Gene 1.0 ST Array 10923161 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21457335 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361776 - -
          RatRef-12 Expression BeadChip ILMN_58090 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline