Search
Search

Fam178b| Gene Symbol: Fam178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244842.2 | XP_006244904.1 | 15 - 16 | 2029 | 57 | |
| XM_006244843.2 | XP_006244905.1 | 16 - 17 | 2143 | 57 | ||
| XM_006244844.2 | XP_006244906.1 | 16 - 17 | 2100 | 57 | ||
| XM_006244846.2 | XP_006244908.1 | 5 - 6 | 1548 | 57 | ||
| XM_006244847.2 | XP_006244909.1 | 6 - 7 | 1378 | 57 | ||
| XM_008767041.1 | XP_008765263.1 | 14 - 15 | 1775 | 57 | ||
Set Membership: |
|