Search
Search

MGC94335| Gene Symbol: MGC94335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004251.1 | NP_001004251.1 | 3 - 4 | 980 | 153 | |
| GenBank mRNA | BC079246.1 | - | 3 - 4 | 980 | 7132862 | 153 |
Set Membership: |
|