Search
Search
| Gene Symbol: Tpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031137.1 | NP_112399.1 | 633 | 67 | ||
| XM_006244852.2 | XP_006244914.1 | 1088 | 67 | |||
| XM_006244853.2 | XP_006244915.1 | 1088 | 67 | |||
| XM_006244870.2 | XP_006244932.1 | 5 - 6 | 1088 | 67 | ||
| GenBank mRNA | U50194.1 | - | 633 | 67 | ||
Set Membership: |
|