Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438026_m1
          See other GULP1 GE Assays ›
          Gene Symbol:
          Gulp1
          Gene Name:
          GULP, engulfment adaptor PTB domain containing 1
          Gene Aliases:
          CED-6
          Chromosome Location:
          Chr.9: 51263622 - 51538066 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438026_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gulp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013171.2 NP_001013189.1 10 - 11 951 134
          XM_006244804.2 XP_006244866.1 9 - 10 893 134
          XM_008767023.1 XP_008765245.1 9 - 10 895 134
          GenBank mRNA BC087053.1 - 10 - 11 951 7123670 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000587.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gulp1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s)

          lipid transport
          phagocytosis, engulfment
          apoptotic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P147029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572001 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154659 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131298 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68389 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126674 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154343 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52253 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152134 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81916 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136824 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149852 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42193 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7796 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139694 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174317 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26183 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109292 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164807 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91632 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49099 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147029 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147029 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P147029 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P117270 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11796 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10783 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23096 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84877 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25216 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96546 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36092 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7254 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45965 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 213 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10700 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41701 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80090 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10923038 - -
          Affymetrix Rat Gene 1.0 ST Array 10923048 - -
          Affymetrix Rat Gene 1.0 ST Array 10923049 - -
          Affymetrix Rat Gene 1.0 ST Array 10923040 - -
          Affymetrix Rat Gene 1.0 ST Array 10923041 - -
          Affymetrix Rat Gene 1.0 ST Array 10923042 - -
          Affymetrix Rat Gene 1.0 ST Array 10923043 - -
          Affymetrix Rat Gene 1.0 ST Array 10923037 - -
          Affymetrix Rat Gene 1.0 ST Array 10923047 - -
          Affymetrix Rat Gene 1.0 ST Array 10923039 - -
          Affymetrix Rat Gene 1.0 ST Array 10923044 - -
          Affymetrix Rat Gene 1.0 ST Array 10923045 - -
          Affymetrix Rat Gene 1.1 ST Array 10923036 - -
          Rat Genome 230 2.0 Array 1376285_PM_at - -
          Rat Genome 230 2.0 Array 1376285_at - -
          Affymetrix HT Rat Focus Array 1376285_at - -
          Affymetrix HT Rat Focus Array 1376285_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900004762.rn.1 TC0900000587.rn.1 RTA TC
          Rat Expression Set 230 1376285_at - -
          Rat Expression Set 230 1376285_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10923036 - -
          Rat Genome U34 Set rc_AI639037_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21194956 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352711 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline