Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438284_m1
          See other AOX2 GE Assays ›
          Gene Symbol:
          Aox2
          Gene Name:
          aldehyde oxidase 2
          Gene Aliases:
          Aox2p, Aox3l1
          Chromosome Location:
          Chr.9: 65175452 - 65266496 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438284_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aox2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008522.1 NP_001008522.1 34 - 35 3986 124
          XM_008767106.1 XP_008765328.1 35 - 36 5615 124
          GenBank mRNA AY665588.1 - 34 - 35 3986 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000700.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aox2

          Gene Ontology Categories:

          Function(s) Process(es)

          oxidation-reduction process
          aldehyde oxidase activity
          iron ion binding
          UDP-N-acetylmuramate dehydrogenase activity
          electron carrier activity
          flavin adenine dinucleotide binding
          NAD binding
          2 iron, 2 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P251045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836206 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10230 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119706 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95412 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113157 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65618 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12739 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127825 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101674 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P251045 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P251045 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P251045 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P251045 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96083 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30412 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40002 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68839 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34965 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60371 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19103 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101330 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10923529 - -
          Affymetrix Rat Gene 1.0 ST Array 10923547 - -
          Affymetrix Rat Gene 1.0 ST Array 10923527 - -
          Affymetrix Rat Gene 1.0 ST Array 10923542 - -
          Affymetrix Rat Gene 1.0 ST Array 10923550 - -
          Affymetrix Rat Gene 1.0 ST Array 10923539 - -
          Affymetrix Rat Gene 1.0 ST Array 10923528 - -
          Affymetrix Rat Gene 1.0 ST Array 10923559 - -
          Affymetrix Rat Gene 1.0 ST Array 10923554 - -
          Affymetrix Rat Gene 1.0 ST Array 10923535 - -
          Affymetrix Rat Gene 1.0 ST Array 10923558 - -
          Affymetrix Rat Gene 1.0 ST Array 10923552 - -
          Affymetrix Rat Gene 1.0 ST Array 10923532 - -
          Affymetrix Rat Gene 1.0 ST Array 10923540 - -
          Affymetrix Rat Gene 1.0 ST Array 10923531 - -
          Affymetrix Rat Gene 1.0 ST Array 10923555 - -
          Affymetrix Rat Gene 1.0 ST Array 10923534 - -
          Affymetrix Rat Gene 1.0 ST Array 10923541 - -
          Affymetrix Rat Gene 1.0 ST Array 10923545 - -
          Affymetrix Rat Gene 1.0 ST Array 10923549 - -
          Affymetrix Rat Gene 1.0 ST Array 10923544 - -
          Affymetrix Rat Gene 1.0 ST Array 10923557 - -
          Affymetrix Rat Gene 1.0 ST Array 10923548 - -
          Affymetrix Rat Gene 1.0 ST Array 10923537 - -
          Affymetrix Rat Gene 1.0 ST Array 10923526 - -
          Affymetrix Rat Gene 1.0 ST Array 10923538 - -
          Affymetrix Rat Gene 1.0 ST Array 10923543 - -
          Affymetrix Rat Gene 1.0 ST Array 10923536 - -
          Affymetrix Rat Gene 1.0 ST Array 10923530 - -
          Affymetrix Rat Gene 1.0 ST Array 10923533 - -
          Affymetrix Rat Gene 1.0 ST Array 10923525 - -
          Affymetrix Rat Gene 1.0 ST Array 10923556 - -
          Affymetrix Rat Gene 1.0 ST Array 10923546 - -
          Affymetrix Rat Gene 1.0 ST Array 10923553 - -
          Affymetrix Rat Gene 1.0 ST Array 10923551 - -
          Affymetrix Rat Gene 1.1 ST Array 10923524 - -
          Affymetrix Rat Clariom S Assay PSR0900005687.rn.1 TC0900000700.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900005689.rn.1 TC0900000700.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10923524 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20865730 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376937 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline