Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438459_m1
          See other MORN5 GE Assays ›
          Gene Symbol:
          Morn5
          Gene Name:
          MORN repeat containing 5
          Gene Aliases:
          LRRGT00041, RGD1307739
          Chromosome Location:
          Chr.3: 15379109 - 15407252 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438459_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Morn5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047912.1 NP_001041377.1 4 - 5 423 71
          GenBank mRNA AY383696.1 - 4 - 5 423 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000316.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P102477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98257 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158031 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25515 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114257 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P102477 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P102477 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P102477 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P102477 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68668 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83356 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46191 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93802 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10835806 - -
          Affymetrix Rat Gene 1.0 ST Array 10835804 - -
          Affymetrix Rat Gene 1.0 ST Array 10835805 - -
          Affymetrix Rat Gene 1.0 ST Array 10835811 - -
          Affymetrix Rat Gene 1.0 ST Array 10835802 - -
          Affymetrix Rat Gene 1.0 ST Array 10835807 - -
          Affymetrix Rat Gene 1.0 ST Array 10835801 - -
          Affymetrix Rat Gene 1.0 ST Array 10835809 - -
          Affymetrix Rat Gene 1.0 ST Array 10835810 - -
          Affymetrix Rat Gene 1.0 ST Array 10835808 - -
          Affymetrix Rat Gene 1.1 ST Array 10835800 - -
          Rat Genome 230 2.0 Array 1393893_at - -
          Rat Genome 230 2.0 Array 1393893_PM_at - -
          Affymetrix HT Rat Focus Array 1393893_PM_at - -
          Affymetrix HT Rat Focus Array 1393893_at - -
          Affymetrix Rat Clariom S Assay PSR0300003499.rn.1 TC0300000316.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300003495.rn.1 TC0300000316.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300003497.rn.1 TC0300000316.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300003496.rn.1 TC0300000316.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300003500.rn.1 TC0300000316.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300003498.rn.1 TC0300000316.rn.1 RTA TC
          Rat Expression Set 230 1393893_PM_at - -
          Rat Expression Set 230 1393893_at - -
          Affymetrix Rat Gene 1.0 ST Array 10835800 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline