Search
Search
| Gene Symbol: Psmd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106569.2 | NP_001100039.2 | 8 - 9 | 1140 | 66 | |
| XM_006233969.2 | XP_006234031.1 | 8 - 9 | 1669 | 66 | ||
| XM_006233970.2 | XP_006234032.1 | 9 - 10 | 1759 | 66 | ||
| GenBank mRNA | BC166850.1 | - | 8 - 9 | 1122 | 9088649 | 66 |
Set Membership: |
|