Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438566_m1
          See other GLE1 GE Assays ›
          Gene Symbol:
          Gle1
          Gene Name:
          GLE1 RNA export mediator
          Gene Aliases:
          Gle1l
          Chromosome Location:
          Chr.3: 8498086 - 8530212 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438566_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gle1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025731.1 NP_001020902.1 2 - 3 381 97
          XM_006233836.2 XP_006233898.1 2 - 3 417 97
          XM_006233838.2 XP_006233900.1 2 - 3 416 97
          GenBank mRNA BC099088.1 - 2 - 3 381 7456179 97
          FQ217346.1 - 2 - 3 402 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000193.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gle1

          Panther Classification:

          Molecular Function -

          translation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          protein transport
          poly(A)+ mRNA export from nucleus
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1007333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86901 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1007333 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1007333 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1007333 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1007333 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41223 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72202 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14148 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10835045 - -
          Affymetrix Rat Gene 1.0 ST Array 10835039 - -
          Affymetrix Rat Gene 1.0 ST Array 10835042 - -
          Affymetrix Rat Gene 1.0 ST Array 10835050 - -
          Affymetrix Rat Gene 1.0 ST Array 10835043 - -
          Affymetrix Rat Gene 1.0 ST Array 10835049 - -
          Affymetrix Rat Gene 1.0 ST Array 10835048 - -
          Affymetrix Rat Gene 1.0 ST Array 10835052 - -
          Affymetrix Rat Gene 1.0 ST Array 10835053 - -
          Affymetrix Rat Gene 1.0 ST Array 10835047 - -
          Affymetrix Rat Gene 1.0 ST Array 10835040 - -
          Affymetrix Rat Gene 1.0 ST Array 10835044 - -
          Affymetrix Rat Gene 1.0 ST Array 10835041 - -
          Affymetrix Rat Gene 1.0 ST Array 10835046 - -
          Affymetrix Rat Gene 1.0 ST Array 10835038 - -
          Affymetrix Rat Gene 1.0 ST Array 10835051 - -
          Affymetrix Rat Gene 1.1 ST Array 10835037 - -
          Rat Genome 230 2.0 Array 1374690_at - -
          Rat Genome 230 2.0 Array 1393357_PM_at - -
          Rat Genome 230 2.0 Array 1374690_PM_at - -
          Rat Genome 230 2.0 Array 1393357_at - -
          Affymetrix HT Rat Focus Array 1393357_PM_at - -
          Affymetrix HT Rat Focus Array 1393357_at - -
          Affymetrix Rat Clariom S Assay PSR0300002052.rn.1 TC0300000193.rn.1 RTA TC
          Rat Expression Set 230 1374690_at - -
          Rat Expression Set 230 1374690_PM_at - -
          Rat Expression Set 230 1393357_at - -
          Rat Expression Set 230 1393357_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10835037 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348859 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline