Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438607_m1
          See other NDUFA8 GE Assays ›
          Gene Symbol:
          Ndufa8
          Gene Name:
          NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8
          Gene Aliases:
          Aa2-258
          Chromosome Location:
          Chr.3: 15362888 - 15379391 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438607_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufa8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047862.1 NP_001041327.1 3 - 4 308 51
          XM_008761741.1 XP_008759963.1 2 - 3 336 51
          GenBank mRNA AY325167.1 - 3 - 4 308 51
          FQ226210.1 - 1 - 2 170 51
          FQ230936.1 - 1 - 2 170 51
          FQ234165.1 - 1 - 2 170 51

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002947.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P423528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57232 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81399 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140598 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P423528 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P423528 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P423528 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P423528 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90932 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61375 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10844731 - -
          Affymetrix Rat Gene 1.0 ST Array 10844729 - -
          Affymetrix Rat Gene 1.0 ST Array 10844728 - -
          Affymetrix Rat Gene 1.0 ST Array 10844727 - -
          Affymetrix Rat Gene 1.0 ST Array 10844730 - -
          Affymetrix Rat Gene 1.1 ST Array 10844726 - -
          Rat Genome 230 2.0 Array 1371355_at - -
          Rat Genome 230 2.0 Array 1371355_PM_at - -
          Affymetrix HT Rat Focus Array 1371355_PM_at - -
          Affymetrix HT Rat Focus Array 1371355_at - -
          Affymetrix Rat Clariom S Assay PSR0300022994.rn.1 TC0300002947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300022996.rn.1 TC0300002947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300022992.rn.1 TC0300002947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300022989.rn.1 TC0300002947.rn.1 RTA TC
          Rat Expression Set 230 1371355_at - -
          Rat Expression Set 230 1371355_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10844726 - -
          Rat Genome U34 Set rc_AI232012_at - -
          Rat Genome U34 Set rc_AI232012_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21561914 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371540 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline