Search
Search
| Gene Symbol: Ncs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024366.1 | NP_077342.1 | 5 - 6 | 458 | 78 | |
| XM_006233925.2 | XP_006233987.1 | 5 - 6 | 762 | 78 | ||
| XM_008761679.1 | XP_008759901.1 | 4 - 5 | 353 | 78 | ||
| GenBank mRNA | L27421.1 | - | 5 - 6 | 398 | 78 | |
| X82188.1 | - | 5 - 6 | 458 | 78 | ||
Set Membership: |
|