Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438680_m1
          See other MAPKAP1 GE Assays ›
          Gene Symbol:
          Mapkap1
          Gene Name:
          mitogen-activated protein kinase associated protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 13484952 - 13701772 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438680_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mapkap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011964.1 NP_001011964.1 11 - 12 1886 58
          XM_006233949.2 XP_006234011.1 12 - 13 2072 58
          XM_006233950.2 XP_006234012.1 11 - 12 1624 58
          XM_008761720.1 XP_008759942.1 10 - 11 1563 58
          XM_008761721.1 XP_008759943.1 10 - 11 1586 58
          XM_008761722.1 XP_008759944.1 9 - 10 1496 58
          XM_008761723.1 XP_008759945.1 9 - 10 1174 58
          GenBank mRNA BC079073.1 - 11 - 12 1886 7122012 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000286.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mapkap1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          substantia nigra development
          negative regulation of Ras protein signal transduction
          molecular_function
          phosphatidylinositol-4,5-bisphosphate binding
          phosphatidylinositol-3,4,5-trisphosphate binding
          Ras GTPase binding
          protein kinase binding
          phosphatidylinositol-3,4-bisphosphate binding
          phosphatidic acid binding
          phosphatidylinositol-3,5-bisphosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1009178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100021 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14800 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134598 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140491 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171317 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52564 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56514 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174308 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43264 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85475 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48878 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34307 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136314 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17367 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107693 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49476 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77855 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13874 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1009178 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1009178 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1009178 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1009178 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45953 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104870 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79084 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57768 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68373 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68491 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8477 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50083 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61675 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83261 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70260 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28346 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6092 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10835675 - -
          Affymetrix Rat Gene 1.0 ST Array 10835671 - -
          Affymetrix Rat Gene 1.0 ST Array 10835672 - -
          Affymetrix Rat Gene 1.0 ST Array 10835674 - -
          Affymetrix Rat Gene 1.0 ST Array 10835667 - -
          Affymetrix Rat Gene 1.0 ST Array 10835676 - -
          Affymetrix Rat Gene 1.0 ST Array 10835670 - -
          Affymetrix Rat Gene 1.0 ST Array 10835673 - -
          Affymetrix Rat Gene 1.0 ST Array 10835668 - -
          Affymetrix Rat Gene 1.0 ST Array 10835669 - -
          Affymetrix Rat Gene 1.0 ST Array 10835677 - -
          Affymetrix Rat Gene 1.1 ST Array 10835666 - -
          Rat Genome 230 2.0 Array 1389167_PM_at - -
          Rat Genome 230 2.0 Array 1377007_PM_at - -
          Rat Genome 230 2.0 Array 1389167_at - -
          Rat Genome 230 2.0 Array 1377007_at - -
          Affymetrix HT Rat Focus Array 1389167_at - -
          Affymetrix HT Rat Focus Array 1389167_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300003238.rn.1 TC0300000286.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300003237.rn.1 TC0300000286.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300003236.rn.1 TC0300000286.rn.1 RTA TC
          Rat Expression Set 230 1389167_PM_at - -
          Rat Expression Set 230 1389167_at - -
          Affymetrix Rat Gene 1.0 ST Array 10835666 - -
          Rat Genome U34 Set rc_AI069970_s_at - -
          Rat Genome U34 Set rc_AI228279_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22221065 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376703 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline