Search
Search
| Gene Symbol: Npdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004231.1 | NP_001004231.1 | 3 - 4 | 465 | 80 | |
| XM_006233584.2 | XP_006233646.1 | 2 - 3 | 1103 | 80 | ||
| GenBank mRNA | BC079156.1 | - | 3 - 4 | 465 | 7128989 | 80 |
| FQ214366.1 | - | 3 - 4 | 309 | 80 | ||
Set Membership: |
|