Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01438901_m1
          See other PSD4 GE Assays ›
          Gene Symbol:
          Psd4
          Gene Name:
          pleckstrin and Sec7 domain containing 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 1478431 - 1515098 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01438901_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134881.1 NP_001128353.1 18 - 19 3216 70
          XM_006233591.2 XP_006233653.1 16 - 17 3299 70
          XM_006233592.2 XP_006233654.1 16 - 17 3084 70
          XM_006233593.2 XP_006233655.1 16 - 17 3290 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000018.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of ARF protein signal transduction
          positive regulation of GTPase activity
          ARF guanyl-nucleotide exchange factor activity
          phospholipid binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P127727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516907 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39910 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110123 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120808 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7702 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127727 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127727 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P127727 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P562505 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P119289 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10834130 - -
          Affymetrix Rat Gene 1.0 ST Array 10834117 - -
          Affymetrix Rat Gene 1.0 ST Array 10834134 - -
          Affymetrix Rat Gene 1.0 ST Array 10834121 - -
          Affymetrix Rat Gene 1.0 ST Array 10834132 - -
          Affymetrix Rat Gene 1.0 ST Array 10834118 - -
          Affymetrix Rat Gene 1.0 ST Array 10834124 - -
          Affymetrix Rat Gene 1.0 ST Array 10834120 - -
          Affymetrix Rat Gene 1.0 ST Array 10834116 - -
          Affymetrix Rat Gene 1.0 ST Array 10834127 - -
          Affymetrix Rat Gene 1.0 ST Array 10834125 - -
          Affymetrix Rat Gene 1.0 ST Array 10834128 - -
          Affymetrix Rat Gene 1.0 ST Array 10834119 - -
          Affymetrix Rat Gene 1.0 ST Array 10834133 - -
          Affymetrix Rat Gene 1.0 ST Array 10834122 - -
          Affymetrix Rat Gene 1.0 ST Array 10834129 - -
          Affymetrix Rat Gene 1.0 ST Array 10834135 - -
          Affymetrix Rat Gene 1.0 ST Array 10834131 - -
          Affymetrix Rat Gene 1.0 ST Array 10834126 - -
          Affymetrix Rat Gene 1.1 ST Array 10834115 - -
          Rat Genome 230 2.0 Array 1390195_at - -
          Rat Genome 230 2.0 Array 1390195_PM_at - -
          Affymetrix HT Rat Focus Array 1390195_at - -
          Affymetrix HT Rat Focus Array 1390195_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300000095.rn.1 TC0300000018.rn.1 RTA TC
          Rat Expression Set 230 1390195_at - -
          Rat Expression Set 230 1390195_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10834115 - -
          Rat Genome U34 Set rc_AI029511_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369661 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline