Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01439129_s1
          See other MRRF GE Assays ›
          Gene Symbol:
          Mrrf
          Gene Name:
          mitochondrial ribosome recycling factor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 15463919 - 15520621 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01439129_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrrf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008354.1 NP_001008355.1 7 1556 137
          XM_006234068.1 XP_006234130.1 8 1543 137
          XM_006234069.1 XP_006234131.1 6 1400 137
          GenBank mRNA BC085779.1 - 7 1556 7106905 137

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrrf

          Panther Classification:

          Molecular Function -

          translation factor translation release factor translational protein

          Gene Ontology Categories:

          Function(s)

          translation
          ribosome disassembly

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P050091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811402 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31982 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54634 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124868 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38447 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P567088 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P567088 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P567088 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P777199 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P326108 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P567088 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P527536 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46254 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25431 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64496 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60842 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10835814 - -
          Affymetrix Rat Gene 1.0 ST Array 10835815 - -
          Affymetrix Rat Gene 1.0 ST Array 10835816 - -
          Affymetrix Rat Gene 1.1 ST Array 10835813 - -
          Rat Genome 230 2.0 Array 1376415_PM_at - -
          Rat Genome 230 2.0 Array 1376415_at - -
          Rat Genome 230 2.0 Array 1396227_at - -
          Rat Genome 230 2.0 Array 1396227_PM_at - -
          Affymetrix HT Rat Focus Array 1396227_at - -
          Affymetrix HT Rat Focus Array 1396227_PM_at - -
          Affymetrix HT Rat Focus Array 1376415_at - -
          Affymetrix HT Rat Focus Array 1376415_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300003521.rn.1 TC0300000317.rn.1 RTA TC
          Rat Expression Set 230 1376415_at - -
          Rat Expression Set 230 1376415_PM_at - -
          Rat Expression Set 230 1396227_PM_at - -
          Rat Expression Set 230 1396227_at - -
          Affymetrix Rat Gene 1.0 ST Array 10835813 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20727895 - -
          ABI Rat Genome Survey Microarray 21239090 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353840 - -
          RatRef-12 Expression BeadChip ILMN_62534 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0