Search
Search

Ccdc183| Gene Symbol: Ccdc183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025040.2 | NP_001020211.2 | 6 - 7 | 700 | 82 | |
| XM_006233635.1 | XP_006233697.1 | 7 - 8 | 1349 | 82 | ||
| GenBank mRNA | BC160897.1 | - | 6 - 7 | 700 | 7453119 | 82 |
Set Membership: |
|