Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01439336_m1
          See other ROPN1 GE Assays ›
          Gene Symbol:
          Ropn1
          Gene Name:
          rhophilin associated tail protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 69326785 - 69355788 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01439336_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ropn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025628.1 NP_001020799.1 5 - 6 738 99
          XM_006248431.2 XP_006248493.1 5 - 6 681 99
          GenBank mRNA BC099132.1 - 5 - 6 738 7453834 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001716.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ropn1

          Gene Ontology Categories:

          Function(s)

          regulation of protein phosphorylation
          sperm motility
          cilium organization
          sperm capacitation
          protein localization to cilium

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P022283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102093 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111389 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103271 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176902 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022283 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022283 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P022283 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P534812 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8862 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11200 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11797 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10754562 - -
          Affymetrix Rat Gene 1.0 ST Array 10754559 - -
          Affymetrix Rat Gene 1.0 ST Array 10754558 - -
          Affymetrix Rat Gene 1.0 ST Array 10754561 - -
          Affymetrix Rat Gene 1.0 ST Array 10754557 - -
          Affymetrix Rat Gene 1.0 ST Array 10754560 - -
          Affymetrix Rat Gene 1.1 ST Array 10754556 - -
          Affymetrix Rat Clariom S Assay PSR1100010616.rn.1 TC1100001716.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10754556 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21846925 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350446 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline