Search
Search

RGD1310641| Gene Symbol: RGD1310641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079701.1 | NP_001073169.1 | 10 - 11 | 1429 | 95 | |
| XM_006237999.2 | XP_006238061.1 | 10 - 11 | 1486 | 95 | ||
| XM_008763628.1 | XP_008761850.1 | 10 - 11 | 1342 | 95 | ||
| XM_008763629.1 | XP_008761851.1 | 9 - 10 | 1327 | 95 | ||
| GenBank mRNA | BC098003.1 | - | 3 - 4 | 363 | 95 | |
| BC128759.1 | - | 10 - 11 | 1429 | 7112639 | 95 | |
Set Membership: |
|