Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106647.3 | NP_001100117.2 | 3 - 4 | 757 | 89 | |
NM_001256084.1 | NP_001243013.1 | 3 - 4 | 757 | 89 | ||
GenBank mRNA | BC159418.1 | - | 3 - 4 | 757 | 8370352 | 89 |
BF523560.1 | - | 2 - 3 | 167 | 89 | ||
BI280304.1 | - | 4 - 5 | 89 | |||
FQ215132.1 | - | 3 - 4 | 719 | 89 | ||
FQ224976.1 | - | 3 - 4 | 754 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |