Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01439866_g1
          See other DNAJB5 GE Assays ›
          Gene Symbol:
          Dnajb5
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily B, member 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 58393197 - 58402162 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01439866_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajb5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108004.1 NP_001101474.1 4 - 5 1121 73
          XM_006238070.2 XP_006238132.1 4 - 5 1154 73
          XM_006238071.2 XP_006238133.1 3 - 4 1557 73
          XM_006238072.2 XP_006238134.1 3 - 4 973 73
          GenBank mRNA BC166524.1 - 3 - 4 970 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000513.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          protein folding
          response to unfolded protein
          unfolded protein binding
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P331706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22584 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P331706 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P331706 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P331706 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P331706 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P344906 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10868431 - -
          Affymetrix Rat Gene 1.0 ST Array 10868433 - -
          Affymetrix Rat Gene 1.0 ST Array 10868429 - -
          Affymetrix Rat Gene 1.0 ST Array 10868432 - -
          Affymetrix Rat Gene 1.0 ST Array 10868430 - -
          Affymetrix Rat Gene 1.1 ST Array 10868428 - -
          Rat Genome 230 2.0 Array 1372144_at - -
          Rat Genome 230 2.0 Array 1384511_at - -
          Rat Genome 230 2.0 Array 1372144_PM_at - -
          Rat Genome 230 2.0 Array 1384511_PM_at - -
          Rat Genome 230 2.0 Array 1375762_PM_at - -
          Rat Genome 230 2.0 Array 1375762_at - -
          Affymetrix HT Rat Focus Array 1375762_at - -
          Affymetrix HT Rat Focus Array 1372144_at - -
          Affymetrix HT Rat Focus Array 1384511_PM_at - -
          Affymetrix HT Rat Focus Array 1372144_PM_at - -
          Affymetrix HT Rat Focus Array 1384511_at - -
          Affymetrix HT Rat Focus Array 1375762_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500003130.rn.1 TC0500000513.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500003131.rn.1 TC0500000513.rn.1 RTA TC
          Rat Expression Set 230 1375762_PM_at - -
          Rat Expression Set 230 1372144_at - -
          Rat Expression Set 230 1372144_PM_at - -
          Rat Expression Set 230 1375762_at - -
          Rat Expression Set 230 1384511_PM_at - -
          Rat Expression Set 230 1384511_at - -
          Affymetrix Rat Gene 1.0 ST Array 10868428 - -
          Rat Genome U34 Set rc_AA891542_at - -
          Rat Genome U34 Set rc_AI172021_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21742690 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357975 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline