Search
Search
| Gene Symbol: Ahi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002277.1 | NP_001002277.1 | 10 - 11 | 1817 | 64 | |
| GenBank mRNA | AY647140.1 | - | 10 - 11 | 1817 | 64 | |
| FQ226565.1 | - | 2098 | 64 | |||
Set Membership: |
|