Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01440591_m1
          See other GPR126 GE Assays ›
          Gene Symbol:
          Gpr126
          Gene Name:
          G protein-coupled receptor 126
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 8593342 - 8751534 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01440591_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr126
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006227647.2 XP_006227709.1 22 - 23 3848 101
          XM_218313.8 XP_218313.7 23 - 24 3933 101

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P947743 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P947743 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P947743 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P947743 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P947743 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P550537 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10716958 - -
          Affymetrix Rat Gene 1.0 ST Array 10716944 - -
          Affymetrix Rat Gene 1.0 ST Array 10716954 - -
          Affymetrix Rat Gene 1.0 ST Array 10716955 - -
          Affymetrix Rat Gene 1.0 ST Array 10716948 - -
          Affymetrix Rat Gene 1.0 ST Array 10716961 - -
          Affymetrix Rat Gene 1.0 ST Array 10716946 - -
          Affymetrix Rat Gene 1.0 ST Array 10716959 - -
          Affymetrix Rat Gene 1.0 ST Array 10716960 - -
          Affymetrix Rat Gene 1.0 ST Array 10716949 - -
          Affymetrix Rat Gene 1.0 ST Array 10716943 - -
          Affymetrix Rat Gene 1.0 ST Array 10716956 - -
          Affymetrix Rat Gene 1.0 ST Array 10716942 - -
          Affymetrix Rat Gene 1.0 ST Array 10716963 - -
          Affymetrix Rat Gene 1.0 ST Array 10716950 - -
          Affymetrix Rat Gene 1.0 ST Array 10716945 - -
          Affymetrix Rat Gene 1.0 ST Array 10716953 - -
          Affymetrix Rat Gene 1.0 ST Array 10716952 - -
          Affymetrix Rat Gene 1.0 ST Array 10716951 - -
          Affymetrix Rat Gene 1.0 ST Array 10716962 - -
          Affymetrix Rat Gene 1.0 ST Array 10716947 - -
          Affymetrix Rat Gene 1.0 ST Array 10716940 - -
          Affymetrix Rat Gene 1.0 ST Array 10716957 - -
          Affymetrix Rat Gene 1.1 ST Array 10716939 - -
          Rat Genome 230 2.0 Array 1382955_at - -
          Rat Genome 230 2.0 Array 1382955_PM_at - -
          Affymetrix HT Rat Focus Array 1382955_at - -
          Affymetrix HT Rat Focus Array 1382955_PM_at - -
          Rat Expression Set 230 1382955_PM_at - -
          Rat Expression Set 230 1382955_at - -
          Affymetrix Rat Gene 1.0 ST Array 10716939 - -
          Rat Genome U34 Set rc_AA997539_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20904805 - -
          ABI Rat Genome Survey Microarray 21749007 - -
          ABI Rat Genome Survey Microarray 22116762 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375120 - -
          RatRef-12 Expression BeadChip ILMN_65215 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline