Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01440751_m1
          See other FBXO30 GE Assays ›
          Gene Symbol:
          Fbxo30
          Gene Name:
          F-box protein 30
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 5366120 - 5389560 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01440751_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxo30
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007690.1 NP_001007691.1 2 - 3 2201 165
          XM_006227640.2 XP_006227702.1 2 - 3 2243 165
          GenBank mRNA BC083823.1 - 2 - 3 2201 7111792 165

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000059.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxo30

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          ubiquitin-protein transferase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P547213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100132 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53747 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P547213 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P547213 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P547213 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P177845 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97294 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32072 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10701789 - -
          Affymetrix Rat Gene 1.0 ST Array 10701790 - -
          Affymetrix Rat Gene 1.0 ST Array 10701791 - -
          Affymetrix Rat Gene 1.1 ST Array 10701788 - -
          Rat Genome 230 2.0 Array 1392747_PM_at - -
          Rat Genome 230 2.0 Array 1392747_at - -
          Rat Genome 230 2.0 Array 1382059_at - -
          Rat Genome 230 2.0 Array 1382059_PM_at - -
          Affymetrix HT Rat Focus Array 1382059_PM_at - -
          Affymetrix HT Rat Focus Array 1382059_at - -
          Affymetrix Rat Clariom S Assay PSR0100000312.rn.1 TC0100000059.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000314.rn.1 TC0100000059.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000315.rn.1 TC0100000059.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000317.rn.1 TC0100000059.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000313.rn.1 TC0100000059.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000316.rn.1 TC0100000059.rn.1 RTA TC
          Rat Expression Set 230 1382059_at - -
          Rat Expression Set 230 1382059_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10701788 - -
          Rat Genome U34 Set rc_AI639506_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22204864 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650644 - -
          RatRef-12 Expression BeadChip ILMN_52726 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline