Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01440844_m1
          See other AIG1 GE Assays ›
          Gene Symbol:
          Aig1
          Gene Name:
          androgen-induced 1
          Gene Aliases:
          RGD1562920
          Chromosome Location:
          Chr.1: 7816841 - 8038808 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01440844_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aig1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134425.1 NP_001127897.1 1 - 2 168 65
          XM_006227642.2 XP_006227704.1 1 - 2 367 65
          XM_008758602.1 XP_008756824.1 1 - 2 364 65
          XM_008758603.1 XP_008756825.1 1 - 2 366 65
          XM_008758604.1 XP_008756826.1 1 - 2 362 65
          GenBank mRNA BC167098.1 - 1 - 2 168 7319120 65
          FQ228880.1 - 1 - 2 240 65
          FQ229550.1 - 1 - 2 185 65
          FQ229983.1 - 1 - 2 184 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004029.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11403 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152013 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139462 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50158 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116376 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92053 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115990 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25302 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113116 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68092 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71272 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142695 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136254 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7435 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122048 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12542 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144781 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10008 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10008 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10008 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17239 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10716928 - -
          Affymetrix Rat Gene 1.0 ST Array 10716931 - -
          Affymetrix Rat Gene 1.0 ST Array 10716926 - -
          Affymetrix Rat Gene 1.0 ST Array 10716932 - -
          Affymetrix Rat Gene 1.0 ST Array 10716929 - -
          Affymetrix Rat Gene 1.0 ST Array 10716930 - -
          Affymetrix Rat Gene 1.1 ST Array 10716925 - -
          Rat Genome 230 2.0 Array 1375845_PM_at - -
          Rat Genome 230 2.0 Array 1375845_at - -
          Rat Genome 230 2.0 Array 1381824_PM_at - -
          Rat Genome 230 2.0 Array 1381824_at - -
          Affymetrix HT Rat Focus Array 1381824_PM_at - -
          Affymetrix HT Rat Focus Array 1375845_PM_at - -
          Affymetrix HT Rat Focus Array 1381824_at - -
          Affymetrix HT Rat Focus Array 1375845_at - -
          Affymetrix Rat Clariom S Assay PSR0100032771.rn.1 TC0100004029.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032774.rn.1 TC0100004029.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032777.rn.1 TC0100004029.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032773.rn.1 TC0100004029.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032775.rn.1 TC0100004029.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032776.rn.1 TC0100004029.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032770.rn.1 TC0100004029.rn.1 RTA TC
          Rat Expression Set 230 1375845_PM_at - -
          Rat Expression Set 230 1375845_at - -
          Rat Expression Set 230 1381824_PM_at - -
          Rat Expression Set 230 1381824_at - -
          Affymetrix Rat Gene 1.0 ST Array 10716925 - -
          Rat Genome U34 Set rc_AA925603_at - -
          Rat Genome U34 Set rc_AI231309_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20994687 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360255 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline