Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01440906_m1
          See other SESN1 GE Assays ›
          Gene Symbol:
          Sesn1
          Gene Name:
          sestrin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 46666983 - 46758306 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01440906_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sesn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106396.1 NP_001099866.1 2 - 3 866 63
          XM_006256560.2 XP_006256622.1 4 - 5 1604 63
          XM_006256561.2 XP_006256623.1 4 - 5 616 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000836.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s)

          regulation of protein kinase B signaling
          regulation of response to reactive oxygen species

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P389571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79950 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175828 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173655 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96796 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170389 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12933 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 659 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104409 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98234 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143072 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120699 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P389571 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P389571 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P389571 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17837 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48067 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74071 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11127 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55883 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63945 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71509 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10830568 - -
          Affymetrix Rat Gene 1.0 ST Array 10830572 - -
          Affymetrix Rat Gene 1.0 ST Array 10830570 - -
          Affymetrix Rat Gene 1.0 ST Array 10830571 - -
          Affymetrix Rat Gene 1.0 ST Array 10830569 - -
          Affymetrix Rat Gene 1.0 ST Array 10830566 - -
          Affymetrix Rat Gene 1.0 ST Array 10830567 - -
          Affymetrix Rat Gene 1.0 ST Array 10830573 - -
          Affymetrix Rat Gene 1.0 ST Array 10830562 - -
          Affymetrix Rat Gene 1.1 ST Array 10830561 - -
          Rat Genome 230 2.0 Array 1372248_PM_at - -
          Rat Genome 230 2.0 Array 1395809_at - -
          Rat Genome 230 2.0 Array 1372248_at - -
          Rat Genome 230 2.0 Array 1395809_PM_at - -
          Affymetrix HT Rat Focus Array 1395809_at - -
          Affymetrix HT Rat Focus Array 1395809_PM_at - -
          Affymetrix HT Rat Focus Array 1372248_PM_at - -
          Affymetrix HT Rat Focus Array 1372248_at - -
          Affymetrix Rat Clariom S Assay PSR2000007047.rn.1 TC2000000836.rn.1 RTA TC
          Rat Expression Set 230 1372248_at - -
          Rat Expression Set 230 1372248_PM_at - -
          Rat Expression Set 230 1395809_PM_at - -
          Rat Expression Set 230 1395809_at - -
          Affymetrix Rat Gene 1.0 ST Array 10830561 - -
          Rat Genome U34 Set rc_AI137579_at - -
          Rat Genome U34 Set rc_AI104331_at - -
          Rat Genome U34 Set rc_AI137622_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20798554 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353298 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline