Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01441080_g1
          See other QRSL1 GE Assays ›
          Gene Symbol:
          Qrsl1
          Gene Name:
          glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 48855839 - 48881110 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01441080_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Qrsl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014034.1 NP_001014056.1 8 - 9 1101 92
          XM_006256588.2 XP_006256650.1 8 - 9 1140 92
          GenBank mRNA BC089213.1 - 8 - 9 1101 7302358 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001745.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Qrsl1

          Panther Classification:

          Molecular Function -

          ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial translation
          glutaminyl-tRNAGln biosynthesis via transamidation
          ATP binding
          glutaminyl-tRNA synthase (glutamine-hydrolyzing) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P715389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149824 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P715389 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P715389 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P715389 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P511202 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P715389 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96115 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28172 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10833847 - -
          Affymetrix Rat Gene 1.0 ST Array 10833844 - -
          Affymetrix Rat Gene 1.0 ST Array 10833846 - -
          Affymetrix Rat Gene 1.0 ST Array 10833848 - -
          Affymetrix Rat Gene 1.0 ST Array 10833845 - -
          Affymetrix Rat Gene 1.0 ST Array 10833838 - -
          Affymetrix Rat Gene 1.0 ST Array 10833842 - -
          Affymetrix Rat Gene 1.0 ST Array 10833841 - -
          Affymetrix Rat Gene 1.0 ST Array 10833839 - -
          Affymetrix Rat Gene 1.0 ST Array 10833843 - -
          Affymetrix Rat Gene 1.0 ST Array 10833840 - -
          Affymetrix Rat Gene 1.1 ST Array 10833837 - -
          Rat Genome 230 2.0 Array 1383831_PM_at - -
          Rat Genome 230 2.0 Array 1383831_at - -
          Rat Genome 230 2.0 Array 1380842_at - -
          Rat Genome 230 2.0 Array 1380842_PM_at - -
          Affymetrix HT Rat Focus Array 1380842_at - -
          Affymetrix HT Rat Focus Array 1383831_PM_at - -
          Affymetrix HT Rat Focus Array 1380842_PM_at - -
          Affymetrix HT Rat Focus Array 1383831_at - -
          Affymetrix Rat Clariom S Assay PSR2000014083.rn.1 TC2000001745.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014085.rn.1 TC2000001745.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014087.rn.1 TC2000001745.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014084.rn.1 TC2000001745.rn.1 RTA TC
          Rat Expression Set 230 1380842_PM_at - -
          Rat Expression Set 230 1380842_at - -
          Rat Expression Set 230 1383831_PM_at - -
          Rat Expression Set 230 1383831_at - -
          Affymetrix Rat Gene 1.0 ST Array 10833837 - -
          Rat Genome U34 Set rc_AA997173_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21903147 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650024 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline