Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01441351_g1
          See other SUMO1 GE Assays ›
          Gene Symbol:
          Sumo1
          Gene Name:
          small ubiquitin-like modifier 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 66453428 - 66483614 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01441351_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sumo1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009672.1 NP_001009672.1 4 - 5 276 105
          GenBank mRNA BC088322.1 - 4 - 5 276 7325996 105
          FQ211636.1 - 4 - 5 313 105
          FQ214927.1 - 4 - 5 346 105
          FQ215001.1 - 4 - 5 356 105
          FQ215643.1 - 4 - 5 105
          FQ234204.1 - 4 - 5 348 105

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sumo1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          protein sumoylation
          PML body organization
          positive regulation of protein complex assembly
          regulation of protein stability
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of transcription, DNA-templated
          palate development
          regulation of cardiac muscle cell contraction
          protein localization to nuclear pore
          positive regulation of calcium-transporting ATPase activity
          regulation of calcium ion transmembrane transport
          protein binding
          transcription factor binding
          SUMO transferase activity
          ubiquitin protein ligase binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P253858 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P135903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737602 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62157 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41407 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176310 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P135903 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P253858 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P135903 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P253858 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P135903 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P253858 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P253858 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35267 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73314 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10928426 - -
          Affymetrix Rat Gene 1.0 ST Array 10928425 - -
          Affymetrix Rat Gene 1.0 ST Array 10928424 - -
          Affymetrix Rat Gene 1.1 ST Array 10928423 - -
          Rat Genome 230 2.0 Array 1399095_at - -
          Rat Genome 230 2.0 Array 1399095_PM_at - -
          Affymetrix HT Rat Focus Array 1399095_PM_at - -
          Affymetrix HT Rat Focus Array 1399095_at - -
          Affymetrix Rat Clariom S Assay PSR0900017018.rn.1 TC0900002051.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900017019.rn.1 TC0900002051.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900017020.rn.1 TC0900002051.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900017017.rn.1 TC0900002051.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900017021.rn.1 TC0900002051.rn.1 RTA TC
          Rat Expression Set 230 1399095_PM_at - -
          Rat Expression Set 230 1399095_at - -
          Affymetrix Rat Gene 1.0 ST Array 10928423 - -
          Rat Genome U34 Set AA686579_at - -
          Rat Genome U34 Set rc_AI059527_f_at - -
          Rat Genome U34 Set rc_AA850550_at - -
          Rat Genome U34 Set rc_AI059527_r_at - -
          Rat Genome U34 Set rc_AI059527_i_at - -
          Rat Genome U34 Set AA686579_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21133082 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362579 - -
          RatRef-12 Expression BeadChip ILMN_61563 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline