Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01441525_gH
          See other PRSS39 GE Assays ›
          Gene Symbol:
          Prss39
          Gene Name:
          protease, serine, 39
          Gene Aliases:
          Prss40, Tesp1, Tesp2
          Chromosome Location:
          Chr.9: 41006606 - 41012607 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01441525_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss39
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013226.1 NP_001013244.1 2 - 3 639 86
          XM_006244717.2 XP_006244779.1 3 - 4 695 86
          XM_006244718.2 XP_006244780.1 3 - 4 323 86
          GenBank mRNA BC079254.1 - 2 - 3 639 7133201 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000452.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prss39

          Panther Classification:

          Molecular Function -

          serine protease

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P043708 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P136576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113555 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P136576 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043708 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043708 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P136576 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P043708 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P136576 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19385 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10922448 - -
          Affymetrix Rat Gene 1.0 ST Array 10922447 - -
          Affymetrix Rat Gene 1.0 ST Array 10922445 - -
          Affymetrix Rat Gene 1.0 ST Array 10922446 - -
          Affymetrix Rat Gene 1.0 ST Array 10922444 - -
          Affymetrix Rat Gene 1.1 ST Array 10922443 - -
          Rat Genome 230 2.0 Array 1384713_PM_at - -
          Rat Genome 230 2.0 Array 1384713_at - -
          Rat Genome 230 2.0 Array 1393850_PM_at - -
          Rat Genome 230 2.0 Array 1393850_at - -
          Affymetrix Rat Clariom S Assay PSR0900003536.rn.1 TC0900000452.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900003537.rn.1 TC0900000452.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900003535.rn.1 TC0900000452.rn.1 RTA TC
          Rat Expression Set 230 1384713_at - -
          Rat Expression Set 230 1384713_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10922443 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21875625 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372836 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline