Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01441815_m1
          See other FGFR4 GE Assays ›
          Gene Symbol:
          Fgfr4
          Gene Name:
          fibroblast growth factor receptor 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 9990072 - 10004335 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01441815_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgfr4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109904.1 NP_001103374.1 5 - 6 767 59
          XM_006253606.2 XP_006253668.1 5 - 6 781 59
          XM_008771485.1 XP_008769707.1 5 - 6 781 59
          GenBank mRNA BC100260.1 - 4 - 5 859 7100540 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001332.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          organ induction
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          positive regulation of gene expression
          regulation of extracellular matrix disassembly
          cell migration
          peptidyl-tyrosine phosphorylation
          regulation of lipid metabolic process
          lung development
          glucose homeostasis
          positive regulation of catalytic activity
          positive regulation of proteolysis
          protein autophosphorylation
          phosphate ion homeostasis
          alveolar secondary septum development
          positive regulation of ERK1 and ERK2 cascade
          regulation of bile acid biosynthetic process
          response to bile acid
          regulation of cholesterol homeostasis
          positive regulation of DNA biosynthetic process
          fibroblast growth factor-activated receptor activity
          ATP binding
          heparin binding
          fibroblast growth factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P456689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27158 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145474 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P456689 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P456689 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P456689 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P456689 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701188388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74121 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61247 - -
          Agilent Rat cDNA Microarray (G4105A) 701188388 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797326 - -
          Affymetrix Rat Gene 1.0 ST Array 10797341 - -
          Affymetrix Rat Gene 1.0 ST Array 10797336 - -
          Affymetrix Rat Gene 1.0 ST Array 10797337 - -
          Affymetrix Rat Gene 1.0 ST Array 10797338 - -
          Affymetrix Rat Gene 1.0 ST Array 10797339 - -
          Affymetrix Rat Gene 1.0 ST Array 10797325 - -
          Affymetrix Rat Gene 1.0 ST Array 10797340 - -
          Affymetrix Rat Gene 1.0 ST Array 10797334 - -
          Affymetrix Rat Gene 1.0 ST Array 10797335 - -
          Affymetrix Rat Gene 1.0 ST Array 10797329 - -
          Affymetrix Rat Gene 1.0 ST Array 10797343 - -
          Affymetrix Rat Gene 1.0 ST Array 10797330 - -
          Affymetrix Rat Gene 1.0 ST Array 10797342 - -
          Affymetrix Rat Gene 1.0 ST Array 10797331 - -
          Affymetrix Rat Gene 1.0 ST Array 10797324 - -
          Affymetrix Rat Gene 1.0 ST Array 10797328 - -
          Affymetrix Rat Gene 1.0 ST Array 10797327 - -
          Affymetrix Rat Gene 1.0 ST Array 10797332 - -
          Affymetrix Rat Gene 1.1 ST Array 10797323 - -
          Rat Genome 230 2.0 Array 1389801_PM_at - -
          Rat Genome 230 2.0 Array 1389801_at - -
          Affymetrix HT Rat Focus Array 1389801_at - -
          Affymetrix HT Rat Focus Array 1389801_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700009255.rn.1 TC1700001332.rn.1 RTA TC
          Rat Expression Set 230 1389801_PM_at - -
          Rat Expression Set 230 1389801_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797323 - -
          Rat Genome U34 Set M91599mRNA_g_at - -
          Rat Genome U34 Set M91599mRNA_at - -
          Rat Genome U34 Set rc_AI175948_s_at - -
          Rat Genome U34 Set M91599mRNA_at - -
          Rat Genome U34 Set M91599mRNA_g_at - -
          Rat Genome U34 Set rc_AI175948_s_at - -
          Rat Genome U34 Set M91599mRNA_at - -
          Rat Genome U34 Set M91599mRNA_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21189725 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364341 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline