Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442032_g1
          See other RNF44 GE Assays ›
          Gene Symbol:
          Rnf44
          Gene Name:
          ring finger protein 44
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 10458029 - 10472410 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442032_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf44
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024795.1 NP_001019966.1 9 - 10 1349 61
          XM_006253633.2 XP_006253695.1 9 - 10 2346 61
          XM_006253634.2 XP_006253696.1 9 - 10 1339 61
          XM_006253637.2 XP_006253699.1 9 - 10 2868 61
          XM_006253641.2 XP_006253703.1 9 - 10 2282 61
          XM_006253642.2 XP_006253704.1 9 - 10 2804 61
          XM_006253644.2 XP_006253706.1 9 - 10 1275 61
          XM_008771498.1 XP_008769720.1 9 - 10 2865 61
          GenBank mRNA BC098030.1 - 9 - 10 1349 7456296 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000185.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf44

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P475795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698260 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P475795 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P475795 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P475795 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P475795 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10794147 - -
          Affymetrix Rat Gene 1.0 ST Array 10794156 - -
          Affymetrix Rat Gene 1.0 ST Array 10794155 - -
          Affymetrix Rat Gene 1.0 ST Array 10794151 - -
          Affymetrix Rat Gene 1.0 ST Array 10794150 - -
          Affymetrix Rat Gene 1.0 ST Array 10794154 - -
          Affymetrix Rat Gene 1.0 ST Array 10794157 - -
          Affymetrix Rat Gene 1.0 ST Array 10794158 - -
          Affymetrix Rat Gene 1.0 ST Array 10794152 - -
          Affymetrix Rat Gene 1.0 ST Array 10794148 - -
          Affymetrix Rat Gene 1.0 ST Array 10794153 - -
          Affymetrix Rat Gene 1.1 ST Array 10794146 - -
          Rat Genome 230 2.0 Array 1373936_at - -
          Rat Genome 230 2.0 Array 1373936_PM_at - -
          Rat Genome 230 2.0 Array 1383919_at - -
          Rat Genome 230 2.0 Array 1383919_PM_at - -
          Affymetrix HT Rat Focus Array 1373936_PM_at - -
          Affymetrix HT Rat Focus Array 1373936_at - -
          Affymetrix Rat Clariom S Assay PSR1700001558.rn.1 TC1700000185.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700001556.rn.1 TC1700000185.rn.1 RTA TC
          Rat Expression Set 230 1373936_at - -
          Rat Expression Set 230 1373936_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10794146 - -
          Rat Genome U34 Set rc_AA946475_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20796934 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359873 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline