Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442156_g1
          See other FASTK GE Assays ›
          Gene Symbol:
          Fastk
          Gene Name:
          Fas-activated serine/threonine kinase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 7260529 - 7264590 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442156_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fastk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006235892.2 XP_006235954.1 6 - 7 1289 67
          XM_006235894.2 XP_006235956.1 6 - 7 1276 67
          XM_006235895.1 XP_006235957.1 6 - 7 1273 67
          XM_006235896.2 XP_006235958.1 6 - 7 1290 67
          GenBank mRNA BC083715.1 - 6 - 7 1293 7190690 67
          FQ215763.1 - 7 - 8 67
          FQ228972.1 - 6 - 7 1229 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000109.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          regulation of RNA splicing
          protein serine/threonine kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P729801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142945 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P729801 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P729801 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P729801 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P484485 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852897 - -
          Affymetrix Rat Gene 1.0 ST Array 10852906 - -
          Affymetrix Rat Gene 1.0 ST Array 10852899 - -
          Affymetrix Rat Gene 1.0 ST Array 10852900 - -
          Affymetrix Rat Gene 1.0 ST Array 10852901 - -
          Affymetrix Rat Gene 1.0 ST Array 10852898 - -
          Affymetrix Rat Gene 1.0 ST Array 10852902 - -
          Affymetrix Rat Gene 1.0 ST Array 10852903 - -
          Affymetrix Rat Gene 1.1 ST Array 10852896 - -
          Rat Genome 230 2.0 Array 1372551_PM_at - -
          Rat Genome 230 2.0 Array 1372551_at - -
          Affymetrix HT Rat Focus Array 1372551_PM_at - -
          Affymetrix HT Rat Focus Array 1372551_at - -
          Affymetrix Rat Clariom S Assay PSR0400000773.rn.1 TC0400000109.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400000775.rn.1 TC0400000109.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400000774.rn.1 TC0400000109.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400000781.rn.1 TC0400000109.rn.1 RTA TC
          Rat Expression Set 230 1372551_at - -
          Rat Expression Set 230 1372551_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10852896 - -
          Rat Genome U34 Set rc_AI227743_at - -
          Rat Genome U34 Set rc_AI104285_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22129815 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376279 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline