Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442194_m1
          See other CCDC146 GE Assays ›
          Gene Symbol:
          Ccdc146
          Gene Name:
          coiled-coil domain containing 146
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 10269798 - 10421724 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442194_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc146
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025044.1 NP_001020215.1 15 - 16 2605 96
          GenBank mRNA BC082003.1 - 15 - 16 2605 7131395 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000163.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc146

          Panther Classification:

          Molecular Function -

          structural protein

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P154811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97152 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85929 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32269 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86818 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119341 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P154811 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P154811 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P154811 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15071 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18809 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32234 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30273 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24706 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56959 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10853155 - -
          Affymetrix Rat Gene 1.0 ST Array 10853163 - -
          Affymetrix Rat Gene 1.0 ST Array 10853158 - -
          Affymetrix Rat Gene 1.0 ST Array 10853157 - -
          Affymetrix Rat Gene 1.0 ST Array 10853169 - -
          Affymetrix Rat Gene 1.0 ST Array 10853162 - -
          Affymetrix Rat Gene 1.0 ST Array 10853154 - -
          Affymetrix Rat Gene 1.0 ST Array 10853165 - -
          Affymetrix Rat Gene 1.0 ST Array 10853168 - -
          Affymetrix Rat Gene 1.0 ST Array 10853170 - -
          Affymetrix Rat Gene 1.0 ST Array 10853161 - -
          Affymetrix Rat Gene 1.0 ST Array 10853166 - -
          Affymetrix Rat Gene 1.0 ST Array 10853160 - -
          Affymetrix Rat Gene 1.0 ST Array 10853167 - -
          Affymetrix Rat Gene 1.0 ST Array 10853156 - -
          Affymetrix Rat Gene 1.0 ST Array 10853164 - -
          Affymetrix Rat Gene 1.0 ST Array 10853159 - -
          Affymetrix Rat Gene 1.0 ST Array 10853153 - -
          Affymetrix Rat Gene 1.1 ST Array 10853152 - -
          Affymetrix Rat Clariom S Assay PSR0400001280.rn.1 TC0400000163.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10853152 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22329123 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365511 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline