Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442493_m1
          See other NUPL2 GE Assays ›
          Gene Symbol:
          Nupl2
          Gene Name:
          nucleoporin like 2
          Gene Aliases:
          RGD1560934
          Chromosome Location:
          Chr.4: 7512326 - 7529106 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442493_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nupl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109217.1 NP_001102687.1 2 - 3 463 92
          GenBank mRNA FQ227080.1 - 2 - 3 429 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002190.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transmembrane transport
          porin activity
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P606085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179661 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125423 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P606085 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P606085 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P606085 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P606085 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10860106 - -
          Affymetrix Rat Gene 1.0 ST Array 10860105 - -
          Affymetrix Rat Gene 1.0 ST Array 10860110 - -
          Affymetrix Rat Gene 1.0 ST Array 10860109 - -
          Affymetrix Rat Gene 1.0 ST Array 10860108 - -
          Affymetrix Rat Gene 1.0 ST Array 10860107 - -
          Affymetrix Rat Gene 1.0 ST Array 10860104 - -
          Affymetrix Rat Gene 1.1 ST Array 10860103 - -
          Rat Genome 230 2.0 Array 1390212_at - -
          Rat Genome 230 2.0 Array 1390212_PM_at - -
          Affymetrix HT Rat Focus Array 1390212_PM_at - -
          Affymetrix HT Rat Focus Array 1390212_at - -
          Affymetrix Rat Clariom S Assay PSR0400015654.rn.1 TC0400002190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015652.rn.1 TC0400002190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015647.rn.1 TC0400002190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015657.rn.1 TC0400002190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015650.rn.1 TC0400002190.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400015649.rn.1 TC0400002190.rn.1 RTA TC
          Rat Expression Set 230 1390212_PM_at - -
          Rat Expression Set 230 1390212_at - -
          Affymetrix Rat Gene 1.0 ST Array 10860103 - -
          Rat Genome U34 Set rc_AI010156_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21593547 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373608 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline