Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01442522_m1
          See other KCNH2 GE Assays ›
          Gene Symbol:
          Kcnh2
          Gene Name:
          potassium channel, voltage gated eag related subfamily H, member 2
          Gene Aliases:
          ERG1
          Chromosome Location:
          Chr.4: 7355161 - 7387243 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01442522_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnh2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053949.1 NP_446401.1 14 - 15 3341 62
          XM_006235867.2 XP_006235929.1 14 - 15 3474 62
          XM_008762630.1 XP_008760852.1 10 - 11 2662 62
          GenBank mRNA AY669863.1 - 10 - 11 2315 62
          Z96106.1 - 14 - 15 3341 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000114.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnh2

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphorelay signal transduction system
          potassium ion transport
          brain development
          spinal cord development
          signal transduction by protein phosphorylation
          regulation of ion transmembrane transport
          cellular response to drug
          regulation of membrane potential
          protein heterooligomerization
          potassium ion homeostasis
          cardiac muscle contraction
          regulation of membrane repolarization
          regulation of ventricular cardiac muscle cell membrane repolarization
          potassium ion export
          potassium ion transmembrane transport
          ventricular cardiac muscle cell action potential
          membrane depolarization during action potential
          membrane repolarization during action potential
          membrane repolarization during cardiac muscle cell action potential
          regulation of heart rate by cardiac conduction
          regulation of potassium ion transmembrane transport
          negative regulation of potassium ion transmembrane transport
          positive regulation of potassium ion transmembrane transport
          negative regulation of potassium ion export
          phosphorelay sensor kinase activity
          ion channel activity
          inward rectifier potassium channel activity
          voltage-gated potassium channel activity
          delayed rectifier potassium channel activity
          potassium channel activity
          protein binding
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity
          scaffold protein binding
          voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P066081 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307405 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22213 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103140 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34218 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P066081 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12992 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12992 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P066081 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P066081 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12992 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12992 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702124116 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27178 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39116 - -
          Agilent Rat cDNA Microarray (G4105A) 702124116 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852944 - -
          Affymetrix Rat Gene 1.0 ST Array 10852948 - -
          Affymetrix Rat Gene 1.0 ST Array 10852946 - -
          Affymetrix Rat Gene 1.0 ST Array 10852947 - -
          Affymetrix Rat Gene 1.0 ST Array 10852951 - -
          Affymetrix Rat Gene 1.0 ST Array 10852935 - -
          Affymetrix Rat Gene 1.0 ST Array 10852936 - -
          Affymetrix Rat Gene 1.0 ST Array 10852941 - -
          Affymetrix Rat Gene 1.0 ST Array 10852939 - -
          Affymetrix Rat Gene 1.0 ST Array 10852937 - -
          Affymetrix Rat Gene 1.0 ST Array 10852949 - -
          Affymetrix Rat Gene 1.0 ST Array 10852940 - -
          Affymetrix Rat Gene 1.0 ST Array 10852942 - -
          Affymetrix Rat Gene 1.0 ST Array 10852938 - -
          Affymetrix Rat Gene 1.0 ST Array 10852950 - -
          Affymetrix Rat Gene 1.0 ST Array 10852952 - -
          Affymetrix Rat Gene 1.0 ST Array 10852943 - -
          Affymetrix Rat Gene 1.1 ST Array 10852934 - -
          Rat Genome 230 2.0 Array 1368343_PM_at - -
          Rat Genome 230 2.0 Array 1368343_at - -
          Affymetrix HT Rat Focus Array 1368343_at - -
          Affymetrix HT Rat Focus Array 1368343_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400000855.rn.1 TC0400000114.rn.1 RTA TC
          Rat Expression Set 230 1368343_at - -
          Rat Expression Set 230 1368343_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10852934 - -
          Rat Genome U34 Set Z96106_at - -
          Rat Genome U34 Set U75210_s_at - -
          Rat Genome U34 Set U75210_s_at - -
          Rat Genome U34 Set Z96106_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21848019 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370853 - -
          RatRef-12 Expression BeadChip ILMN_57973 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline