Search
Search
| Gene Symbol: Ccne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108656.1 | NP_001102126.1 | 7 - 8 | 715 | 85 | |
| XM_006237873.1 | XP_006237935.1 | 7 - 8 | 1259 | 85 | ||
| XM_006237874.1 | XP_006237936.1 | 7 - 8 | 722 | 85 | ||
| XM_006237875.1 | XP_006237937.1 | 7 - 8 | 725 | 85 | ||
Set Membership: |
|